Learn More
Leica Microsystems Kendall Healthcare Replacement Heater and Element for Bacti-Cinerator III Sterilizer
Catalog No. 144892
Change view
Click to view available options
For Use With (Equipment):
Bacti-Cinerator III Sterilizer
Catalog No. For Use With (Equipment)
14-489-2 Bacti-Cinerator III Sterilizer
1 options
This item is not returnable. View return policy
Catalog No. 14-489-2 Supplier Leica Microsystems Supplier No. 39001304
Add to Cart
Add to Cart
This item is not returnable. View return policy
For 14-489.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Bacti-cinerator III sterilizer
For Use With (Equipment) Bacti-Cinerator III Sterilizer
Temperature (English) 1500°F
Temperature (Metric) 815°C
Sterilization Time 5 to 7 sec.