Learn More
AdipoGen Kendomycin
SDP
Catalog No. 502562160
Encompass_Preferred
Chemical. CAS 183202-73-5. Formula C29H42O6. MW 486.6. Isolated from Streptomyces violaceoruber. Antibiotic. Potent endothelin receptor antagonist. Anti-osteoporotic. Antibacterial. Cytotoxic. Mediates its cytotoxic effects, at least in part, through proteasome inhibition.
Catalog No. 50-256-2160 Supplier AdipoGen Supplier No. BVT0001C500
Chemical. CAS 183202-73-5. Formula C29H42O6. MW 486.6. Isolated from Streptomyces violaceoruber. Antibiotic. Potent endothelin receptor antagonist. Anti-osteoporotic. Antibacterial. Cytotoxic. Mediates its cytotoxic effects, at least in part, through proteasome inhibition.