Learn More
Butler Schein Animal Health Kennelsol Disinfectant 1 Gallon
Catalog No. NC1752128
Encompass
KennelSol is a broad spectrum germicidal cleaner, deodorizer and disinfectant formulated for animal care facilities. The concentrated formula knocks out pathogens in one easy labor saving step. This very popular quaternary disinfectant is EPA registered and kills a broad spectrum of pathogens, including many important to the animal care industry. Effective against viruses (including Canine Parvovirus), bacteria, and fungi. KennelSol retains its efficacy even in hard water. KennelSol’s pH neutrality at a dilution of 2 oz. per gallon of water (8 oz. per gallon for canine parvovirus) provides for the highest level of safety for animals and humans with no rinsing necessary. It is non-corrosive and non-staining to surfaces and clothing if spilled. An abundant level of detergent and odor control makes it an extraordinary cleaner while its wintergreen fragrance leaves behind a “fresh and clean” scent.
This item is not returnable. View return policy
Catalog No. NC1752128 Supplier Butler Schein Animal Health Supplier No. 020668
KennelSol is a broad spectrum germicidal cleaner, deodorizer and disinfectant formulated for animal care facilities. The concentrated formula knocks out pathogens in one easy labor saving step. This very popular quaternary disinfectant is EPA registered and kills a broad spectrum of pathogens, including many important to the animal care industry. Effective against viruses (including Canine Parvovirus), bacteria, and fungi. KennelSol retains its efficacy even in hard water. KennelSol’s pH neutrality at a dilution of 2 oz. per gallon of water (8 oz. per gallon for canine parvovirus) provides for the highest level of safety for animals and humans with no rinsing necessary. It is non-corrosive and non-staining to surfaces and clothing if spilled. An abundant level of detergent and odor control makes it an extraordinary cleaner while its wintergreen fragrance leaves behind a “fresh and clean” scent.