Learn More
TARGETMOL CHEMICALS INC KENPAULLONE 25MG
SDP
Catalog No. 502580949
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KenPaullone (9-Bromopaullone) a potent CDK1 CDK2 and CDK5 inhibitor as new enhancer for iTreg cell differentiation. Kenpaullone promotes iTreg cell differentiation through increased and prolonged transcription of foxp3 gene by enhancing TGF(beta)-Smad3 signaling pathway. purity: 98%
Catalog No. 50-258-0949 Supplier TARGETMOL CHEMICALS INC Supplier No. T224725MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KenPaullone (9-Bromopaullone) a potent CDK1 CDK2 and CDK5 inhibitor as new enhancer for iTreg cell differentiation. Kenpaullone promotes iTreg cell differentiation through increased and prolonged transcription of foxp3 gene by enhancing TGF(beta)-Smad3 signaling pathway. purity: 98%