Learn More
Enterprise Technology Solutions Kensington Headphone,Noise-Canceling,Hi-Fi,w/6' Cord,USB-C,BK
SDP
Catalog No. 5000740151
Encompass_Preferred
Designed with sound clarity and device compatibility in mind, the USB-C Hi-Fi Headphones are a high-quality combo that is perfect for e-learning, working from home, educational and office environments and more. Maximum output of 94 decibels is for safe listening and protects ears from hearing loss associated with excessive volume. Powerful, 40mm drivers enable a wider dynamic range to deliver crystal-clear audio. Designed to fit heads of all ages, the adjustable headband is padded. Foam ear pads are wrapped in a soft leatherette cover to provide hours of comfortable use. Headphones are compatible with all USB-C devices running Windows, macOS, iPadOS, Android and Chrome OS.
Catalog No. 50-007-40151 Supplier Enterprise Technology Solutions Supplier No. KMW97456
Designed with sound clarity and device compatibility in mind, the USB-C Hi-Fi Headphones are a high-quality combo that is perfect for e-learning, working from home, educational and office environments and more. Maximum output of 94 decibels is for safe listening and protects ears from hearing loss associated with excessive volume. Powerful, 40mm drivers enable a wider dynamic range to deliver crystal-clear audio. Designed to fit heads of all ages, the adjustable headband is padded. Foam ear pads are wrapped in a soft leatherette cover to provide hours of comfortable use. Headphones are compatible with all USB-C devices running Windows, macOS, iPadOS, Android and Chrome OS.