Learn More
Enterprise Technology Solutions Kensington Mount Stand,f/Webcams/Mics/Lights,w/C-Clamp,20"to30",BK
SDP
Catalog No. 5000740565
Encompass_Preferred
A1000 Telescoping C-Clamp Mount is a professional desktop mount for microphones, webcams and lighting systems. Mount includes 3/8", 1/4" and 5/8" adaptors. Adjustable height stand extends 20" to 30" to accommodate standard mounting equipment. 360-degree swivel mount provides flexible webcam or lighting angles. Cable management clips keep your work space organized. C-clamp attaches to the side of desks and platforms ranging from 1/2" to 3-1/2" thick for stable support and more desk space. Use stand with Kensington Webcam and Ring Light (none included) for a professional video setup. It meets MIL-STD-810H Method 504.3 Contamination by Fluids using disinfectants without degradation.
Catalog No. 50-007-40565 Supplier Enterprise Technology Solutions Supplier No. KMW87654
A1000 Telescoping C-Clamp Mount is a professional desktop mount for microphones, webcams and lighting systems. Mount includes 3/8", 1/4" and 5/8" adaptors. Adjustable height stand extends 20" to 30" to accommodate standard mounting equipment. 360-degree swivel mount provides flexible webcam or lighting angles. Cable management clips keep your work space organized. C-clamp attaches to the side of desks and platforms ranging from 1/2" to 3-1/2" thick for stable support and more desk space. Use stand with Kensington Webcam and Ring Light (none included) for a professional video setup. It meets MIL-STD-810H Method 504.3 Contamination by Fluids using disinfectants without degradation.