Learn More
Enterprise Technology Solutions Kensington Privacy Screen, f/ 24" 16:9 Monitor,24-1/20"x14-3/4"x1/5",BK
SDP
Catalog No. 5000740152
Encompass_Preferred
Monitor privacy screen features a magnetic strip to provide a damage-free way to prevent others from viewing your screen. Limited viewing angle narrows the field of vision to around 30 degrees to help keep the information on your screen private and reduce the chances of a visual data breach. Blue light reduction filters out harmful rays by up to 22 percent to ease eyestrain and reduce the chances of blue light interrupting natural sleep patterns. Low reflective coating reduces glare from outside light sources for improved viewing clarity. Reversible viewing has a matte finish on one side to reduce glare and fingerprints. The other side is glossy to provide a clearer view of your screen.
Catalog No. 50-007-40152 Supplier Enterprise Technology Solutions Supplier No. KMWK58357WW
Monitor privacy screen features a magnetic strip to provide a damage-free way to prevent others from viewing your screen. Limited viewing angle narrows the field of vision to around 30 degrees to help keep the information on your screen private and reduce the chances of a visual data breach. Blue light reduction filters out harmful rays by up to 22 percent to ease eyestrain and reduce the chances of blue light interrupting natural sleep patterns. Low reflective coating reduces glare from outside light sources for improved viewing clarity. Reversible viewing has a matte finish on one side to reduce glare and fingerprints. The other side is glossy to provide a clearer view of your screen.