Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KER047 succinate | 2416307-25-8 | 98.9% | 593.65 | 25 MG
SDP

Catalog No. 5000351465
Encompass_Preferred

KER047 succinate is a potent activin receptor-like kinase-2 (ALK2) inhibitor. It is utilized in the research of metabolic diseases, including fibrodysplasia ossificans progressiva (FOP). This product is intended for research use only and is not sold to patients.

  • Potent activin receptor-like kinase-2 (ALK2) inhibitor
  • Used in research for metabolic diseases like fibrodysplasia ossificans progressiva (FOP)
  • For research use only
  • Not sold to patients
  • Appears as a solid, light yellow to yellow in color
  • Ships at room temperature (continental US)
  • Store at 4°C, sealed, away from moisture
  • Soluble in DMSO (2 mg/mL)
  • Solubility protocols available for in vivo dissolution

Catalog No. 50-003-51465 Supplier Medchemexpress LLC Supplier No. 25MGHY136773A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

KER047 succinate is a potent activin receptor-like kinase-2 (ALK2) inhibitor. It is utilized in the research of metabolic diseases, including fibrodysplasia ossificans progressiva (FOP). This product is intended for research use only and is not sold to patients.

  • Potent activin receptor-like kinase-2 (ALK2) inhibitor
  • Used in research for metabolic diseases like fibrodysplasia ossificans progressiva (FOP)
  • For research use only
  • Not sold to patients
  • Appears as a solid, light yellow to yellow in color
  • Ships at room temperature (continental US)
  • Store at 4°C, sealed, away from moisture
  • Soluble in DMSO (2 mg/mL)
  • Solubility protocols available for in vivo dissolution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.