Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Keracyanin chloride | 18719-76-1 | MFCD00135404 | 99.9% | 630.98 | C27H31ClO15 | 1 MG
SDP

Catalog No. 5000242947
Encompass_Preferred

Keracyanin chloride (Cyanidin 3-rutinoside chloride) is a high-purity anthocyanin research compound with antioxidant, anti-inflammatory, and hypoglycemic properties. It is used as a biochemical and pharmacological standard for studies of NF-κB, FAK, and MAPK signaling and for in vitro assays and oral in vivo experiments where appropriate.

  • Purity 99.9%.
  • Molecular weight 630.98 g/mol.
  • Formula C27H31ClO15.
  • Soluble in DMSO (25 mg/mL); ultrasonic recommended.
  • Store sealed, away from moisture; in solution -80°C (6 months) or -20°C (1 month).
  • Common applications: antioxidant assays and signaling pathway studies.

Catalog No. 50-002-42947 Supplier Medchemexpress LLC Supplier No. HY1059351MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Keracyanin chloride (Cyanidin 3-rutinoside chloride) is a high-purity anthocyanin research compound with antioxidant, anti-inflammatory, and hypoglycemic properties. It is used as a biochemical and pharmacological standard for studies of NF-κB, FAK, and MAPK signaling and for in vitro assays and oral in vivo experiments where appropriate.

  • Purity 99.9%.
  • Molecular weight 630.98 g/mol.
  • Formula C27H31ClO15.
  • Soluble in DMSO (25 mg/mL); ultrasonic recommended.
  • Store sealed, away from moisture; in solution -80°C (6 months) or -20°C (1 month).
  • Common applications: antioxidant assays and signaling pathway studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.