Learn More
Medchemexpress LLC Keratin sulfate | 169799-44-4 | 84.2% | 500 MG
SDP
Catalog No. 5000385009
Encompass_Preferred
Keratin sulfate is a heparin-related proteoglycan that interacts with the receptor or alters its stability and function. It does not enhance the mitogenic effect of FGF-2.
  • Heparin-related proteoglycan
  • Interacts with receptors or alters their stability and function
  • Does not enhance the mitogenic effect of FGF-2
  • Solid appearance
  • Off-white to light yellow color
  • Solubility in H2O: 40 mg/mL (needs ultrasonic)
  • For research use only
Catalog No. 50-003-85009 Supplier Medchemexpress LLC Supplier No. HY172443500MG
Keratin sulfate is a heparin-related proteoglycan that interacts with the receptor or alters its stability and function. It does not enhance the mitogenic effect of FGF-2.
  • Heparin-related proteoglycan
  • Interacts with receptors or alters their stability and function
  • Does not enhance the mitogenic effect of FGF-2
  • Solid appearance
  • Off-white to light yellow color
  • Solubility in H2O: 40 mg/mL (needs ultrasonic)
  • For research use only