Learn More
Promocell GmbH PromoCell Keratinocyte Growth Medium 2 500 ml, ready-to-use, with Supplement Mix C-39016 and CaCl 2-Solution C-34005
Catalog No. 50306172
Encompass
PromoCell Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 0.125 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.33 μg / ml || Epinephrine | 0.39 μg / ml || Transferrin (recombinant human) | 10 μg / ml || CaCl2 | 0.06 mM |
Catalog No. 50-306-172 Supplier Promocell GmbH Supplier No. C20011
PromoCell Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 0.125 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.33 μg / ml || Epinephrine | 0.39 μg / ml || Transferrin (recombinant human) | 10 μg / ml || CaCl2 | 0.06 mM |