Learn More
Promocell GmbH PromoCell Keratinocyte Growth Medium 2 KIT 500 ml, Contains: C-20211, C-39011
Catalog No. 50306173
Encompass
PromoCell Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 0.125 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.33 μg / ml || Epinephrine | 0.39 μg / ml || Transferrin (recombinant human) | 10 μg / ml || CaCl2 | 0.06 mM |
Catalog No. 50-306-173 Supplier Promocell GmbH Supplier No. C20111
PromoCell Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.| Media supplement | Final supplement concentration|| Bovine Pituitary Extract | 0.004 ml / ml || Epidermal Growth Factor (recombinant human) | 0.125 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 0.33 μg / ml || Epinephrine | 0.39 μg / ml || Transferrin (recombinant human) | 10 μg / ml || CaCl2 | 0.06 mM |