Learn More
ATCC Keratinocyte Growth Kit
Supplier:  ATCC PCS200040
Encompass
Keratinocyte Growth Kit contains components that when added to Dermal Cell Basal Medium (ATCC PCS-200-030) create a complete ATCC Primary Cell Solutions culture environment for keratinocytes derived from a normal human epidermis (eg, Primary Epidermal Keratinocytes, Normal, Human Neonatal Foreskin).
Catalog No. 50-238-4177
 Restricted Item