Learn More
Sigma Aldrich Fine Chemicals Biosciences Kern PLJ series precision1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7428631EA
Encompass
Range of precision balances with enormous weighing ranges - ideal for heavy tare containers or large samplesFeaturesHigh-quality milligram balance with enormous weighing range up to 2100 g - ideal for large samples or heavy tare containersErgonomically optimised keypad for left and righthanded usersAutomatic internal adjustment guarantees high degree of accuracy and makes the balance independent of its location of use. Ideal for mobile applications which require verification such as ambulatory gold and jewellery purchasingDraught shield standard for models with weighing plate size 80 mm weighing space A˜xH 60x150 mmBacklit LCD display digit height 17 mm
Catalog No. 50-290-8582