Learn More
Crescent Chemical Co Inc KEROSENE LOW ODOR
SDP
Catalog No. 50247779
Encompass_Preferred
Kerosene, low odor Suitable as cooling fluid in horizontal electrophoresis. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 26945.01 CAS 8008-20-6
Catalog No. 50-247-779 Supplier Crescent Chemical Co Inc Supplier No. SE26945.01
Kerosene, low odor Suitable as cooling fluid in horizontal electrophoresis. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 26945.01 CAS 8008-20-6