Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene, Low Odor, Specific Gravity Standard, Reagents

Catalog No. 01334876
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-876 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-876 Supplier Reagents Holdings Llc Supplier No. CK1899001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Kerosene, Low Odor, Specific Gravity Standard, Reagent, use for Specific gravity calibration. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Kerosene, Low Odor, Specific Gravity Standard
Packaging Glass Bottle
CAS 8008-20-6
Specific Gravity 0.767
Physical Form Liquid
Molecular Formula C12H26-C15H32
Boiling Point 175°C
Synonym Paraffin oil
Quantity 1 L
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.