Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene, MilliporeSigma™

Catalog No. 50947992 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle, Pre-Packed
Steel Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
50-947-992 4 L Glass Bottle, Pre-Packed
50-947-630 20 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-947-992 Supplier MilliporeSigma Supplier No. KX00201
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 64742-48-9

Specifications

Boiling Point 180°C to 188°C
Flash Point 54°C
Packaging Glass Bottle, Pre-Packed
Quantity 4 L
Vapor Pressure 0.75 hPa (20°C)
Grade Not Available
Physical Form Liquid
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.