Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene, Reagents

Catalog No. 01337203
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-203 4 L
01-337-204 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-203 Supplier Reagents Holdings Llc Supplier No. C22220004C
Add to Cart
Add to Cart

Kerosene, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 8008-20-6
Molecular Formula C12H26-C15H32
Synonym Paraffin oil

Specifications

Boiling Range 175°C to 325°C
Melting Point -48°C to -26°C
Color Red
Boiling Point 175°C
Melting Range -48°C to -26°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.767
Grade Technical
Physical Form Liquid
Chemical Name or Material Kerosene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.