Learn More
Nielsen Kellerman Kestrel Instruments 2000 Weather Meter, Temperature and Wind, Drop-Tested to MIL-STD-810G, Waterproof to IP67
SDP
Catalog No. NC3944887
Encompass_Preferred
Thanks to the patented external temperature sensor, the Kestrel 2000 weather meter can take accurate water and snow temperature readings, and since it is isolated from the case, you can be sure that the temperature the Kestrel Meter is reading is accurate and not influenced by your hand warming the unit. Forget speculating, know exactly what the conditions are with the Kestrel 2000.
Catalog No. NC3944887 Supplier Nielsen Kellerman Supplier No. 820
Thanks to the patented external temperature sensor, the Kestrel 2000 weather meter can take accurate water and snow temperature readings, and since it is isolated from the case, you can be sure that the temperature the Kestrel Meter is reading is accurate and not influenced by your hand warming the unit. Forget speculating, know exactly what the conditions are with the Kestrel 2000.