Learn More
Nielsen Kellerman Kestrel Instruments 5000 Environmental Meter with Bluetooth Connectivity, Altitude, Pressure, Temperature, Wind, Heat Index, Humidity, Drop-Tested to MIL-STD-810G, Waterproof to IP67
SDP
Catalog No. NC3944890
Encompass_Preferred
The Kestrel 5000 is part of the Kestrel 5 Series platform, which offers a large, high-resolution and contrast display with ease of font readability. It also includes a built-in dual color backlight, extra-strong polycarbonate lens, AA battery operation and optional LiNK iOS and Android wireless connectivity and app support. Like all Kestrel meters, the Kestrel 5000 is drop-proof, dust-proof, waterproof and able to withstand harsh environments without damage – so it can travel with you anywhere you dare to go. Used in firefighting, agriculture, hazardous materials response, meteorology, military, law enforcement, research, education, scientific & mountaineering expeditions and more. Your data log can be transferred to a iOS/Android device wirelessly with the LiNK wireless communication option and Kestrel LiNK app.
Catalog No. NC3944890 Supplier Nielsen Kellerman Supplier No. 0850L
The Kestrel 5000 is part of the Kestrel 5 Series platform, which offers a large, high-resolution and contrast display with ease of font readability. It also includes a built-in dual color backlight, extra-strong polycarbonate lens, AA battery operation and optional LiNK iOS and Android wireless connectivity and app support. Like all Kestrel meters, the Kestrel 5000 is drop-proof, dust-proof, waterproof and able to withstand harsh environments without damage – so it can travel with you anywhere you dare to go. Used in firefighting, agriculture, hazardous materials response, meteorology, military, law enforcement, research, education, scientific & mountaineering expeditions and more. Your data log can be transferred to a iOS/Android device wirelessly with the LiNK wireless communication option and Kestrel LiNK app.