Learn More
Nielsen Kellerman Kestrel Instruments 7000 Environmental Meter, Relative Humidity, Temperature, Heat Index, with Replaceable Sensor, Drop-Tested to MIL-STD-810G, Waterproof to IP67
SDP
Catalog No. NC3944884
Encompass_Preferred
The Kestrel 7000 is the ultimate tool for precise monitoring of relative humidity and temperature. It features a field-replaceable temperature and humidity sensor for easy on-site replacement, reducing downtime. Encased in a rugged, protective shell and backed by a 5-year warranty, this meter withstands harsh conditions. The Kestrel 7000 includes a Certificate of Conformity for accurate readings and offers Bluetooth connectivity, pairing with the Kestrel LiNK app for efficient data management.
Catalog No. NC3944884 Supplier Nielsen Kellerman Supplier No. 870
The Kestrel 7000 is the ultimate tool for precise monitoring of relative humidity and temperature. It features a field-replaceable temperature and humidity sensor for easy on-site replacement, reducing downtime. Encased in a rugged, protective shell and backed by a 5-year warranty, this meter withstands harsh conditions. The Kestrel 7000 includes a Certificate of Conformity for accurate readings and offers Bluetooth connectivity, pairing with the Kestrel LiNK app for efficient data management.