Learn More
Medchemexpress LLC HY-10562 50mg Medchemexpress, Ketanserin CAS:74050-98-9 Purity:>98%
SDP
Catalog No. 501968281
Encompass_Preferred
Medchemexpress, HY-10562 50mg Ketanserin CAS:74050-98-9 Ketanserin is a selective 5-HT receptor antagonist. Ketanserin also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8281 Supplier Medchemexpress LLC Supplier No. HY1056250MG
Medchemexpress, HY-10562 50mg Ketanserin CAS:74050-98-9 Ketanserin is a selective 5-HT receptor antagonist. Ketanserin also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.