Learn More
Selleck Chemical LLC Ketanserin 100mg 74050-98-9 R41468
SDP
Catalog No. 505092
Encompass_Preferred
Ketanserin (R41468) is a specific 5-HT2A serotonin receptor antagonist with Ki of 2.5 nM for rat and human 5-HT2A, used as an antihypertensive drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-509-2 Supplier Selleck Chemical LLC Supplier No. S2232100MG
Ketanserin (R41468) is a specific 5-HT2A serotonin receptor antagonist with Ki of 2.5 nM for rat and human 5-HT2A, used as an antihypertensive drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.