Learn More
AdipoGen Ketanserin tartrate
SDP
Catalog No. 502562396
Encompass_Preferred
Chemical. CAS 83846-83-7. Formula C22H22FN3O3 . C4H6O6. MW 395.4 . 150.1. Selective 5-HT2A serotonin receptor antagonist. ATP-sensitive potassium channel inhibitor. hERG potassium channel blocker.
Catalog No. 50-256-2396 Supplier AdipoGen Supplier No. AGCR10014M010
Chemical. CAS 83846-83-7. Formula C22H22FN3O3 . C4H6O6. MW 395.4 . 150.1. Selective 5-HT2A serotonin receptor antagonist. ATP-sensitive potassium channel inhibitor. hERG potassium channel blocker.