Learn More
Apexbio Technology LLC Ketanserin tartrate 83846-83-7 100mg
SDP
Catalog No. 5000094101
Encompass_Preferred
Ketanserin tartrate (CAS 83846-83-7) is a selective antagonist of 5-HT2 receptors Experimental studies have demonstrated that it inhibits hERG potassium currents in a concentration-dependent manner exhibiting an IC50 of 0 11 M Due to its defined receptor selectivity and electrophysiological properties Ketanserin tartrate is widely employed in pharmacological studies and in research focused on cardiovascular system disorders
Catalog No. 50-000-94101 Supplier Apexbio Technology LLC Supplier No. A4654100
Ketanserin tartrate (CAS 83846-83-7) is a selective antagonist of 5-HT2 receptors Experimental studies have demonstrated that it inhibits hERG potassium currents in a concentration-dependent manner exhibiting an IC50 of 0 11 M Due to its defined receptor selectivity and electrophysiological properties Ketanserin tartrate is widely employed in pharmacological studies and in research focused on cardiovascular system disorders