Learn More
Medchemexpress LLC Ketanserin (tartrate) | 83846-83-7 | 99.9% | 545.51 | 50 MG
SDP
Catalog No. 5000365799
Encompass_Preferred
Ketanserin (R41468) tartrate is a selective 5-HT2 receptor antagonist. It also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM).
  • Identified for use as laboratory chemicals and in the manufacture of substances.
  • Stable under recommended storage conditions.
  • For research use only by experienced personnel.
  • Functions as a 5-HT2A receptor antagonist.
  • Significantly reduces BDNF protein levels in various brain regions.
  • Can significantly reduce BDNF mRNA levels in various brain regions.
  • Inhibits platelet aggregation with an IC50 of 240 nM.
  • Blocks synergistic effects observed for AA with 5-HT, with an IC50 of 152 μM.
  • Inhibits the voltage-dependent step current and tail current of hERG channels within a 5-minute exposure at 0.3 μM.
Catalog No. 50-003-65799 Supplier Medchemexpress LLC Supplier No. 50MGHY10562A
Ketanserin (R41468) tartrate is a selective 5-HT2 receptor antagonist. It also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM).
  • Identified for use as laboratory chemicals and in the manufacture of substances.
  • Stable under recommended storage conditions.
  • For research use only by experienced personnel.
  • Functions as a 5-HT2A receptor antagonist.
  • Significantly reduces BDNF protein levels in various brain regions.
  • Can significantly reduce BDNF mRNA levels in various brain regions.
  • Inhibits platelet aggregation with an IC50 of 240 nM.
  • Blocks synergistic effects observed for AA with 5-HT, with an IC50 of 152 μM.
  • Inhibits the voltage-dependent step current and tail current of hERG channels within a 5-minute exposure at 0.3 μM.