Learn More
Medchemexpress LLC Ketanserin tartrate | 83846-83-7 | 99.9% | 545.51 g/mol | C26H28FN3O9 | 100 MG
SDP
Catalog No. 5000241627
Encompass_Preferred
Ketanserin tartrate is the tartrate salt of ketanserin, a selective serotonin 5-HT2 receptor antagonist used in research to probe 5-HT2 signaling. It additionally blocks cardiac hERG current in a concentration-dependent manner (IC50 = 0.11 μM). The compound is supplied as a high-purity solid intended for research and analytical applications.
  • Selective 5-HT2 receptor antagonist.
  • Blocks hERG current (IC50 = 0.11 μM).
  • High purity (99.9%).
  • Supplied as a solid suitable for analytical use.
  • Molecular weight 545.51 g/mol.
  • Tartrate salt form used in research applications.
Catalog No. 50-002-41627 Supplier Medchemexpress LLC Supplier No. HY10562A100MG
Ketanserin tartrate is the tartrate salt of ketanserin, a selective serotonin 5-HT2 receptor antagonist used in research to probe 5-HT2 signaling. It additionally blocks cardiac hERG current in a concentration-dependent manner (IC50 = 0.11 μM). The compound is supplied as a high-purity solid intended for research and analytical applications.
  • Selective 5-HT2 receptor antagonist.
  • Blocks hERG current (IC50 = 0.11 μM).
  • High purity (99.9%).
  • Supplied as a solid suitable for analytical use.
  • Molecular weight 545.51 g/mol.
  • Tartrate salt form used in research applications.