Learn More
Medchemexpress LLC Ketanserin tartrate | 83846-83-7 | MFCD00084651 | 99.9% | 545.51 g/mol | C26H28FN3O9 | 1 ML
SDP
Catalog No. 5000240169
Encompass_Preferred
Ketanserin tartrate is the tartrate salt of ketanserin, a selective 5-HT2 receptor antagonist that also inhibits hERG current in a concentration-dependent manner. Supplied for research use, it is available as a 10 mM solution in DMSO (1 mL) and as solid quantities, and is accompanied by datasheet, certificate of analysis, and SDS.
Catalog No. 50-002-40169 Supplier Medchemexpress LLC Supplier No. HY10562A10MM 1ML
Ketanserin tartrate is the tartrate salt of ketanserin, a selective 5-HT2 receptor antagonist that also inhibits hERG current in a concentration-dependent manner. Supplied for research use, it is available as a 10 mM solution in DMSO (1 mL) and as solid quantities, and is accompanied by datasheet, certificate of analysis, and SDS.