Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketanserin 98.0+%, TCI America™
SDP

Catalog No. K00511G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K00511G 1 g
K0051200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. K00511G Supplier TCI America Supplier No. K00511G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74050-98-9
Molecular Formula C22H22FN3O3
Molecular Weight (g/mol) 395.434
MDL Number MFCD00083392
InChI Key FPCCSQOGAWCVBH-UHFFFAOYSA-N
Synonym 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,4(1H,3H)-quinazolinedione
PubChem CID 3822
ChEBI CHEBI:6123
IUPAC Name 3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
SMILES C1CN(CCC1C(=O)C2=CC=C(C=C2)F)CCN3C(=O)C4=CC=CC=C4NC3=O

Specifications

Melting Point 233°C
Color White-Yellow
Quantity 1 g
Formula Weight 395.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ketanserin
RTECSNumber VA1411190
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.