Learn More
Cayman Chemical Ketanserin (tartrate), Purity: ≥98%, CAS: 83846-83-7, 100 mg
SDP
Catalog No. 502987192
Encompass_Preferred
A potent 5-HT2 receptor antagonist (IC50 = 6.3 nM; Ki = 2.1 nM); has activity at H1, α1-ARs, and DA receptors; Kis = 10, 10, and 220 nM, respectively); induces dose-dependent inhibition of contractile responses to 5-HT in arteries and veins; significantly decreases BP, BPV, and hypertensive organ damage in spontaneously hypertensive rats
Catalog No. 50-298-7192 Supplier Cayman Chemical Supplier No. 22058100 MG
A potent 5-HT2 receptor antagonist (IC50 = 6.3 nM; Ki = 2.1 nM); has activity at H1, α1-ARs, and DA receptors; Kis = 10, 10, and 220 nM, respectively); induces dose-dependent inhibition of contractile responses to 5-HT in arteries and veins; significantly decreases BP, BPV, and hypertensive organ damage in spontaneously hypertensive rats