Learn More
Medchemexpress LLC Keto-itraconazole | 112560-33-5 | MFCD08063772 | 98.4% | 719.62 g·mol^-1 | C35H36Cl2N8O5 | 5 MG
SDP
Catalog No. 5000227212
Encompass_Preferred
Keto-itraconazole is the primary human metabolite of itraconazole used in research as a potent cytochrome P450 3A (CYP3A) inhibitor. It is supplied as a high-purity solid for in vitro pharmacology, metabolism, and drug-drug interaction studies. Typical small pack sizes are provided for lab-scale experiments.
  • Potent CYP3A inhibitor (unbound IC50 ≈ 4.6 nM in human liver microsomes with midazolam).
  • High purity solid suitable for in vitro assays (reported purity 98.4%).
  • Molecular formula C35H36Cl2N8O5 and molecular weight 719.62 g·mol^-1.
  • Suitable for liver microsome incubations and drug-drug interaction studies.
  • Available in small quantities for research use.
Catalog No. 50-002-27212 Supplier Medchemexpress LLC Supplier No. HY1177895MG
Keto-itraconazole is the primary human metabolite of itraconazole used in research as a potent cytochrome P450 3A (CYP3A) inhibitor. It is supplied as a high-purity solid for in vitro pharmacology, metabolism, and drug-drug interaction studies. Typical small pack sizes are provided for lab-scale experiments.
  • Potent CYP3A inhibitor (unbound IC50 ≈ 4.6 nM in human liver microsomes with midazolam).
  • High purity solid suitable for in vitro assays (reported purity 98.4%).
  • Molecular formula C35H36Cl2N8O5 and molecular weight 719.62 g·mol^-1.
  • Suitable for liver microsome incubations and drug-drug interaction studies.
  • Available in small quantities for research use.