Learn More
Medchemexpress LLC Keto-itraconazole | 112560-33-5 | MFCD08063772 | 98.4% | 719.62 g/mol | C35H36Cl2N8O5 | 1 MG
SDP
Catalog No. 5000227211
Encompass_Preferred
Keto-itraconazole is a metabolic derivative of itraconazole that acts as a potent inhibitor of cytochrome P450 3A (CYP3A). It is provided as an analytical-grade research reagent for in vitro enzyme inhibition and metabolism studies. Molecular formula: C35H36Cl2N8O5; molecular weight: ~719.6 g/mol; typical purity: 98.4%.
  • Potent CYP3A inhibitor with reported unbound IC50 ≈ 4.6 nM.
  • Intended for in vitro enzyme inhibition and metabolism research.
  • High reported purity suitable for analytical experiments.
  • Available in small quantities for screening and mechanistic studies.
  • Provided as a solid; consult the safety data sheet for handling and storage.
Catalog No. 50-002-27211 Supplier Medchemexpress LLC Supplier No. HY1177891MG
Keto-itraconazole is a metabolic derivative of itraconazole that acts as a potent inhibitor of cytochrome P450 3A (CYP3A). It is provided as an analytical-grade research reagent for in vitro enzyme inhibition and metabolism studies. Molecular formula: C35H36Cl2N8O5; molecular weight: ~719.6 g/mol; typical purity: 98.4%.
  • Potent CYP3A inhibitor with reported unbound IC50 ≈ 4.6 nM.
  • Intended for in vitro enzyme inhibition and metabolism research.
  • High reported purity suitable for analytical experiments.
  • Available in small quantities for screening and mechanistic studies.
  • Provided as a solid; consult the safety data sheet for handling and storage.