Learn More
Alkali Scientific Ketoconazole, 1 G
SDP
Catalog No. 5000659867
Encompass_Preferred
Ketoconazole is an antifungal enzyme inhibitor commonly used in fungal research. Available in a 1g format, it is primarily used to study its effects on fungal infections, including dermatophytes and systemic fungal diseases. As an imidazole antifungal, ketoconazole works by inhibiting the synthesis of ergosterol, a critical component of fungal cell membranes. This action leads to the disruption of membrane integrity and ultimately the death of the fungal cell. Ketoconazole is widely used in vitro and in vivo for researching antifungal mechanisms and drug resistance. It is often applied in microbiological studies to evaluate the effectiveness of antifungal agents against common fungal pathogens.
Catalog No. 50-006-59867 Supplier Alkali Scientific Supplier No. KET003CP
Ketoconazole is an antifungal enzyme inhibitor commonly used in fungal research. Available in a 1g format, it is primarily used to study its effects on fungal infections, including dermatophytes and systemic fungal diseases. As an imidazole antifungal, ketoconazole works by inhibiting the synthesis of ergosterol, a critical component of fungal cell membranes. This action leads to the disruption of membrane integrity and ultimately the death of the fungal cell. Ketoconazole is widely used in vitro and in vivo for researching antifungal mechanisms and drug resistance. It is often applied in microbiological studies to evaluate the effectiveness of antifungal agents against common fungal pathogens.