Learn More
Alkali Scientific Ketoconazole, 100 Mg
SDP
Catalog No. 5000659714
Encompass_Preferred
Ketoconazole is a broad-spectrum antifungal agent used in protein synthesis inhibition studies. This 100 mg formulation is often used in research investigating the mechanisms of fungal infections and the impact of antifungal treatments. Ketoconazole works by inhibiting ergosterol synthesis, an essential component of fungal cell membranes, which leads to cell death. It is widely used in studies focusing on fungal cell biology and is also used as a model compound in drug discovery research targeting fungal pathogens. Additionally, ketoconazole has been explored for its anti-inflammatory and anticancer properties.
Catalog No. 50-006-59714 Supplier Alkali Scientific Supplier No. KET001CP
Ketoconazole is a broad-spectrum antifungal agent used in protein synthesis inhibition studies. This 100 mg formulation is often used in research investigating the mechanisms of fungal infections and the impact of antifungal treatments. Ketoconazole works by inhibiting ergosterol synthesis, an essential component of fungal cell membranes, which leads to cell death. It is widely used in studies focusing on fungal cell biology and is also used as a model compound in drug discovery research targeting fungal pathogens. Additionally, ketoconazole has been explored for its anti-inflammatory and anticancer properties.