Learn More
Cayman Chemical Ketoconazole, 65277-42-1, 100 mg
SDP
Supplier:  Cayman Chemical 15212100
Encompass_Preferred
An orally active broad spectrum antifungal agent that blocks ergosterol biosynthesis by inhibiting the fungal CYP isoform CYP51; potently inhibits the mammalian analog CYP51A1 (IC50 = 63.5 nM), as well as a variety of other CYP isoforms
Catalog No. NC0958719