Learn More
Medchemexpress LLC HY-B0105B 10mg Medchemexpress, (-)-Ketoconazole CAS:142128-57-2 Purity:>98%
SDP
Catalog No. 501970096
Encompass_Preferred
Medchemexpress, HY-B0105B 10mg (-)-Ketoconazole CAS:142128-57-2 (-)-Ketoconazole ((-)-R 41400) is one of the enantiomer of Ketoconazole. Ketoconazole is a racemic mixture of two enantiomers, levoketoconazole ((2S,4R)-(−)-ketoconazole) and dextroketoconazole ((2R,4S)-(+)-ketoconazole). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0096 Supplier Medchemexpress LLC Supplier No. HYB0105B10MG
Medchemexpress, HY-B0105B 10mg (-)-Ketoconazole CAS:142128-57-2 (-)-Ketoconazole ((-)-R 41400) is one of the enantiomer of Ketoconazole. Ketoconazole is a racemic mixture of two enantiomers, levoketoconazole ((2S,4R)-(−)-ketoconazole) and dextroketoconazole ((2R,4S)-(+)-ketoconazole). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.