Learn More
Enzo Life Sciences Ketoconazole (50mg). CAS: 65277-42-1
SDP
Supplier:  Enzo Life Sciences BMLEI1070050
Encompass_Preferred
Inhibits cytochrome P-450-dependent steps in the biosynthesis of steroid hormones in vivo. Antimetastatic and antineoplastic activity. Antipsoriatic activity. Orally active 5-lipoxygenase and thromboxane synthase inhibitor. Purity: ≥99% (HPLC). Long Term Storage: Ambient.
Catalog No. 50-201-0551