Learn More
Medchemexpress LLC Ketoconazole | 65277-42-1 | 99.6% | C26H28Cl2N4O4 | 25 MG
SDP
Catalog No. 5000359040
Encompass_Preferred
Ketoconazole (R-41400) is an imidazole anti-fungal agent and an inhibitor of CYP3A4 and CYP24A1. It is for research use only. In studies, it has been observed to inhibit hepatic CYP450 and CYT b5 activities, enhance antiproliferative effects, and promote caspase-independent apoptosis pathways. It is also a potent exosome biogenesis and/or secretion inhibitor.
  • Inhibits hepatic CYP450 and CYT b5 activities
  • Enhances antiproliferative effects and increases systemic calcitriol exposure
  • Promotes activation of caspase-independent apoptosis pathways
  • Potent exosome biogenesis and/or secretion inhibitor
  • Used in histological imaging/staining, cell proliferation/viability assays, and in vivo efficacy studies
  • Assesses lipid peroxidation and cell viability
  • Pre-treats primary mouse hepatocytes to measure PEPCK1 protein levels
Catalog No. 50-003-59040 Supplier Medchemexpress LLC Supplier No. 25MGHYB0105B
Ketoconazole (R-41400) is an imidazole anti-fungal agent and an inhibitor of CYP3A4 and CYP24A1. It is for research use only. In studies, it has been observed to inhibit hepatic CYP450 and CYT b5 activities, enhance antiproliferative effects, and promote caspase-independent apoptosis pathways. It is also a potent exosome biogenesis and/or secretion inhibitor.
  • Inhibits hepatic CYP450 and CYT b5 activities
  • Enhances antiproliferative effects and increases systemic calcitriol exposure
  • Promotes activation of caspase-independent apoptosis pathways
  • Potent exosome biogenesis and/or secretion inhibitor
  • Used in histological imaging/staining, cell proliferation/viability assays, and in vivo efficacy studies
  • Assesses lipid peroxidation and cell viability
  • Pre-treats primary mouse hepatocytes to measure PEPCK1 protein levels