Learn More
Medchemexpress LLC Ketoconazole-d4 | 1398065-75-2 | 99.63% | C26H24D4Cl2N4O4 | 1 MG
SDP
Catalog No. 5000432441
Encompass_Preferred
Ketoconazole-d4 is a deuterium labeled form of Ketoconazole, which is an imidazole anti-fungal agent and a CYP3A4 and CYP24A1 inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Inhibits CYP3A4 and CYP24A1
  • Acts as an anti-fungal agent
  • Suitable for research use only
Catalog No. 50-004-32441 Supplier Medchemexpress LLC Supplier No. HYB0105S11MG
Ketoconazole-d4 is a deuterium labeled form of Ketoconazole, which is an imidazole anti-fungal agent and a CYP3A4 and CYP24A1 inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound
  • Inhibits CYP3A4 and CYP24A1
  • Acts as an anti-fungal agent
  • Suitable for research use only