Learn More
Medchemexpress LLC Ketoconazole-d8 | 1217706-96-1 | 99.8% | C26H20D8Cl2N4O4 | 2.5 MG
SDP
Catalog No. 5000433520
Encompass_Preferred
Ketoconazole-d8 is the deuterium labeled version of Ketoconazole. Ketoconazole is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules and serve as tracers for quantitation during the process of drug development. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Imidazole anti-fungal agent
  • CYP3A4 and CYP24A1 inhibitor
  • Deuterium labeled
Catalog No. 50-004-33520 Supplier Medchemexpress LLC Supplier No. HYB0105S2.5MG
Add to Cart
Add to Cart
Ketoconazole-d8 is the deuterium labeled version of Ketoconazole. Ketoconazole is an imidazole anti-fungal agent, a CYP3A4 and CYP24A1 inhibitor. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules and serve as tracers for quantitation during the process of drug development. Deuteration is notable for its potential to influence the pharmacokinetic and metabolic profiles of drugs. This compound is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Imidazole anti-fungal agent
  • CYP3A4 and CYP24A1 inhibitor
  • Deuterium labeled