Learn More
Selleck Chemical LLC Ketoconazole 100mg 65277-42-1
SDP
Catalog No. 507307
Encompass_Preferred
Ketoconazole inhibits cyclosporine oxidase and testosterone 6 beta-hydroxylase with IC50 of 0.19 mM and 0.22 mM, respectively. Ketoconazole is an androgen biosynthesis inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-730-7 Supplier Selleck Chemical LLC Supplier No. S1353100MG
Ketoconazole inhibits cyclosporine oxidase and testosterone 6 beta-hydroxylase with IC50 of 0.19 mM and 0.22 mM, respectively. Ketoconazole is an androgen biosynthesis inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.