Learn More
Cayman Chemical Ketoconazoled3 GC MS Standards
SDP
Catalog No. 502888318
Encompass_Preferred
An internal standard for the quantification of ketoconazole by GC- or LC-MS
Catalog No. 50-288-8318 Supplier Cayman Chemical Supplier No. 10010656500 UG
An internal standard for the quantification of ketoconazole by GC- or LC-MS