Learn More
Cayman Chemical Ketoconazoled3 GC MS Standards
SDP
Supplier:  Cayman Chemical 100106561 MG
Encompass_Preferred
An internal standard for the quantification of ketoconazole by GC- or LC-MS
Catalog No. 50-288-8316