Learn More
Cayman Chemical Ketoconazole-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1217766-70-5, 5 mg
SDP
Catalog No. 502888317
Encompass_Preferred
An internal standard for the quantification of ketoconazole by GC- or LC-MS
Catalog No. 50-288-8317 Supplier Cayman Chemical Supplier No. 100106565 MG
An internal standard for the quantification of ketoconazole by GC- or LC-MS