Learn More
Med Vet International Ketofen Injection, 100mg/mL, 50mL
Supplier:  Med Vet International RXKETO50
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Kefofen Sterile Solution is recommended for the alleviation of inflammation and pain associated with musculoskeletal disorders in horses. The product is an FDA-approved non-narcotic, nonsteroidal anti-inflammatory agent with analgesic and antipyretic properties. FDA-approved non-narcotic, nonsteroidal anti-inflammatory agent recommended for the alleviation of inflammation and pain associated with musculoskeletal disorders in horses Significantly reduces hoof pain associated with chronic laminitis for up to six hours Showed significant intra-articular anti-inflammatory activity Therapeutically equivalent to flunixin meglumine in the alleviation of pain in carpal model of arthritis NSAID of the propionic acid class that includes ibuprofen, naproxen and fenoprofen Need Syringes & Needles? Click here
Catalog No. 50-282-8711