Learn More
α-Ketoglutaric Acid Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 328-50-7 |
| Flash Point | Not applicable |
| Molecular Formula | C5H6O5 |
| Linear Formula | HOOCCH2CH2COCOOH |
| Quantity | 50 mg |
| MDL Number | MFCD00004165 |
| UN Number | NONH for all modes of transport |
| Synonym | 2-Oxoglutaric Acid; 2-Oxopentanedioic Acid |
| Molecular Weight (g/mol) | 146.10 |
| Formula Weight | 146.10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.