Learn More
Medchemexpress LLC α-ketoglutaric acid (disodium dihydrate) | 1282616-74-3 | MFCD00150702 | 98.0% | 226.09 g/mol | C5H8Na2O7 | 25g
SDP
Catalog No. 5000270355
Encompass_Preferred
-Ketoglutaric acid disodium dihydrate an important dicarboxylic acid is an intermediate in the tricarboxylic acid cycle (TCC) and amino acid metabolism -Ketoglutaric acid disodium dihydrate can be used as building block chemical for the chemical synthesis of heterocycles dietary supplement component of infusion solutions and wound healing compounds or as main component of new elastomers with a wide range of interesting mechanical and chemical properties[1]
Catalog No. 50-002-70355 Supplier Medchemexpress LLC Supplier No. HYW80269925G
-Ketoglutaric acid disodium dihydrate an important dicarboxylic acid is an intermediate in the tricarboxylic acid cycle (TCC) and amino acid metabolism -Ketoglutaric acid disodium dihydrate can be used as building block chemical for the chemical synthesis of heterocycles dietary supplement component of infusion solutions and wound healing compounds or as main component of new elastomers with a wide range of interesting mechanical and chemical properties[1]