Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-ketoglutaric acid (disodium hydrate) | 305-72-6 | MFCD00043347 | 95.0% | 208.08 | C5H6Na2O6 | 1 G
SDP

Catalog No. 5000282149
Encompass_Preferred

α-Ketoglutaric acid (disodium hydrate) is the disodium salt hydrate form of alpha-ketoglutarate, a central metabolite in the tricarboxylic acid (TCA) cycle. Supplied as a white to off-white solid, this reagent is intended for biochemical assays, metabolic studies, and general laboratory research.

  • White to off-white solid appearance.
  • Purity 95.0% (NMR).
  • Molecular formula C5H6Na2O6; molecular weight 208.08.
  • Suitable for biochemical assays and metabolic and enzymatic studies.
  • Storage: 4°C under nitrogen, protected from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small lab pack sizes for research use.

Catalog No. 50-002-82149 Supplier Medchemexpress LLC Supplier No. HYW339545A1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Ketoglutaric acid (disodium hydrate) is the disodium salt hydrate form of alpha-ketoglutarate, a central metabolite in the tricarboxylic acid (TCA) cycle. Supplied as a white to off-white solid, this reagent is intended for biochemical assays, metabolic studies, and general laboratory research.

  • White to off-white solid appearance.
  • Purity 95.0% (NMR).
  • Molecular formula C5H6Na2O6; molecular weight 208.08.
  • Suitable for biochemical assays and metabolic and enzymatic studies.
  • Storage: 4°C under nitrogen, protected from moisture; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small lab pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.