Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Ketoglutaric Acid Disodium Salt, Dihydrate, MP Biomedicals™

Catalog No. ICN15139350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15139350 50 g
02-151-39383 250 g
02-151-39390 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15139350 Supplier MP Biomedicals, Inc Supplier No. 0215139350
Add to Cart
Add to Cart

Chemical Identifiers

CAS 305-72-6
Molecular Formula C5H4Na2O5
Molecular Weight (g/mol) 190.062
MDL Number MFCD00150702
InChI Key YBGBJYVHJTVUSL-UHFFFAOYSA-L
Synonym disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
PubChem CID 31040
IUPAC Name disodium;2-oxopentanedioate
SMILES C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 50 g
Formula Weight 226.1
Physical Form Solid
Chemical Name or Material α-Ketoglutaric Acid Disodium Salt, dihydrate
EINECSNumber 206-167-8
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.