Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Ketoglutaric Acid, MP Biomedicals

Catalog No. ICN201767 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN201767 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN201767 Supplier MP Biomedicals, Inc Supplier No. 0520176705
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 328-50-7
Molecular Formula C5H6O5
Quantity 5 g
MDL Number MFCD00004165
Synonym 2-ketoglutaric acid, alpha-ketoglutaric acid, 2-oxoglutaric acid, 2-oxoglutarate, oxoglutaric acid, alpha-ketoglutarate, 2-ketoglutarate, alphaketoglutaric acid, alpha-oxoglutaric acid, 2-oxo-1,5-pentanedioic acid
InChI Key KPGXRSRHYNQIFN-UHFFFAOYSA-N
SMILES C(CC(=O)O)C(=O)C(=O)O
IUPAC Name 2-oxopentanedioic acid
Molecular Weight (g/mol) 146.098
PubChem CID 51
ChEBI CHEBI:30915
Chemical Name or Material Alpha-Ketoglutaric Acid
Show More Show Less
EINECSNumber 206-330-3
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.